This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287308_g1
See other TMIGD2 GE Assays ›
Gene Symbol:
TMIGD2
Gene Name:
transmembrane and immunoglobulin domain containing 2
Gene Aliases:
CD28H, IGPR-1, IGPR1
Chromosome Location:
Chr.19: 4292227 - 4302439 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287308_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMIGD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308232.1 NP_001295161.1 3 - 4 252 61
NM_144615.2 NP_653216.2 4 - 5 612 61
GenBank mRNA BC015655.1 - 4 - 5 612 4811204 61
HQ452865.1 - 3 - 4 206 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMIGD2

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine production
immunoglobulin production
immune response
signal transduction
T cell costimulation
positive regulation of activated T cell proliferation
positive regulation of angiogenesis
protein binding
coreceptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0