This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287167_m1
See other MYL12A GE Assays ›
Gene Symbol:
MYL12A
Gene Name:
myosin light chain 12A
Gene Aliases:
HEL-S-24, MLC-2B, MLCB, MRCL3, MRLC3, MYL2B
Chromosome Location:
Chr.18: 3247482 - 3256237 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287167_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYL12A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303047.1 NP_001289976.1 2 - 3 463 66
GenBank mRNA BM561281.1 - 2 - 3 180 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001303047.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

muscle contraction
ephrin receptor signaling pathway
platelet aggregation
calcium ion binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0