This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286879_m1
See other KCNT2 GE Assays ›
Gene Symbol:
KCNT2
Gene Name:
potassium sodium-activated channel subfamily T member 2
Gene Aliases:
KCa4.2, SLICK, SLO2.1
Chromosome Location:
Chr.1: 196225779 - 196608905 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286879_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287819.1 NP_001274748.1 2 - 3 293 122
NM_001287820.1 NP_001274749.1 2 - 3 293 122
NM_198503.3 NP_940905.2 2 - 3 293 122
XM_006711294.2 XP_006711357.1 2 - 3 767 122
XM_006711295.2 XP_006711358.1 2 - 3 767 122
XM_011509482.2 XP_011507784.1 2 - 3 767 122
XM_011509483.2 XP_011507785.1 2 - 3 767 122
XM_017001178.1 XP_016856667.1 2 - 3 767 122
XM_017001179.1 XP_016856668.1 2 - 3 767 122
XM_017001180.1 XP_016856669.1 2 - 3 767 122
XM_017001182.1 XP_016856671.1 2 - 3 767 122
XM_017001184.1 XP_016856673.1 2 - 3 767 122
XM_017001185.1 XP_016856674.1 2 - 3 767 122
GenBank mRNA AK131498.1 - 2 - 3 422 122
AK309729.1 - 2 - 3 282 122
AY359444.1 - 2 - 3 231 122
BC103947.1 - 2 - 3 221 122
BC103948.1 - 2 - 3 221 40009146 122
BC103949.1 - 2 - 3 221 40009147 122
BC103950.1 - 2 - 3 221 40009148 122
BX647852.1 - 2 - 3 274 122
DA799907.1 - 2 - 3 293 122
EU251076.1 - 2 - 3 171 122

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC103947.1, XM_017001182.1, XM_017001184.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KCNT2

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transmembrane transport
voltage-gated potassium channel activity
ATP binding
calcium-activated potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0