This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286838_m1
See other PTTG1IP GE Assays ›
Gene Symbol:
PTTG1IP
Gene Name:
pituitary tumor-transforming 1 interacting protein
Gene Aliases:
C21orf1, C21orf3, PBF
Chromosome Location:
Chr.21: 44849585 - 44873903 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286838_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTTG1IP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286822.1 NP_001273751.1 2 - 3 460 86
GenBank mRNA AK298557.1 - 2 - 3 248 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus
positive regulation of protein ubiquitination
negative regulation of DNA damage response, signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of cellular protein catabolic process
p53 binding
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0