This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286825_m1
See other RGS20 GE Assays ›
Gene Symbol:
RGS20
Gene Name:
regulator of G-protein signaling 20
Gene Aliases:
RGSZ1, ZGAP1, g(z)GAP, gz-GAP
Chromosome Location:
Chr.8: 53851808 - 53959304 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286825_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGS20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286674.1 NP_001273603.1 1 - 2 258 82
GenBank mRNA AF366056.1 - 1 - 2 166 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF366056.1, NM_001286674.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
cell death
regulation of G-protein coupled receptor protein signaling pathway
dorsal/ventral axis specification
cell differentiation
regulation of catenin import into nucleus
positive regulation of GTPase activity
cellular response to organic cyclic compound
negative regulation of canonical Wnt signaling pathway
Wnt signaling pathway involved in somitogenesis
GTPase activator activity
protein binding
beta-catenin binding
protein kinase binding
armadillo repeat domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header