This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07286786_m1
See other TMEM9B GE Assays ›
Gene Symbol:
TMEM9B
Gene Name:
TMEM9 domain family member B
Gene Aliases:
C11orf15
Chromosome Location:
Chr.11: 8947201 - 8965011 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07286786_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM9B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286094.1 NP_001273023.1 1 - 2 563 101
GenBank mRNA AK310723.1 - 1 - 2 194 101
AK316216.1 - 1 - 2 194 101
BC015884.1 - 1 - 2 190 3909465 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK316216.1, BC015884.1, NM_001286094.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
signal transducer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0