This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06641546_s1
See other CNP GE Assays ›
Gene Symbol:
CNP
Gene Name:
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases:
CNP1
Chromosome Location:
Chr.17: 41966741 - 41977736 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06641546_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033133.4 NP_149124.3 2 586 77
XM_006721701.3 XP_006721764.1 2 610 77
XM_011524340.2 XP_011522642.1 2 571 77
GenBank mRNA AK026876.1 - 2 543 77
AK098384.1 - 2 532 77
BC001362.2 - 2 523 3051112 77
BC006392.2 - 2 545 4126696 77
BC011046.1 - 2 507 4179083 77
BC028040.1 - 2 547 5248370 77
CB988051.1 - 2 613 77
M19650.1 - 1 395 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CNP

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
chemical synaptic transmission
axonogenesis
aging
adult locomotory behavior
cyclic nucleotide catabolic process
response to toxic substance
substantia nigra development
forebrain development
response to lipopolysaccharide
regulation of mitochondrial membrane permeability
oligodendrocyte differentiation
RNA binding
2',3'-cyclic-nucleotide 3'-phosphodiesterase activity
cyclic nucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0