This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06640309_s1
See other MYO5A GE Assays ›
Gene Symbol:
MYO5A
Gene Name:
myosin VA
Gene Aliases:
GS1, MYH12, MYO5, MYR12
Chromosome Location:
Chr.15: 52307283 - 52529050 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06640309_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYO5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000259.3 NP_000250.3 28 3935 101
NM_001142495.1 NP_001135967.1 28 3935 101
XM_005254397.3 XP_005254454.1 28 3767 101
XM_011521606.2 XP_011519908.2 28 3770 101
XM_011521607.2 XP_011519909.2 28 3770 101
XM_011521608.2 XP_011519910.2 28 3769 101
XM_011521609.2 XP_011519911.2 28 3769 101
XM_011521610.2 XP_011519912.2 28 3770 101
XM_011521611.2 XP_011519913.2 28 3769 101
XM_011521612.2 XP_011519914.2 28 3769 101
XM_017022227.1 XP_016877716.1 27 3662 101
GenBank mRNA AB209487.1 - 21 2548 101
AB209505.1 - 21 2548 101
AB290180.1 - 28 3694 101
AF090424.1 - 1 70 101
AF090425.1 - 1 70 101
AF090426.1 - 1 70 101
AF090427.1 - 1 70 101
AF090428.1 - 1 70 101
S74799.1 - 5 512 101
U90942.1 - 28 3941 101
Y07759.1 - 28 3941 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYO5A

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

transport
exocytosis
post-Golgi vesicle-mediated transport
chemical synaptic transmission
visual perception
vesicle-mediated transport
actin filament-based movement
vesicle transport along actin filament
insulin secretion
melanocyte differentiation
regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
locomotion involved in locomotory behavior
secretory granule localization
melanosome transport
cellular response to insulin stimulus
odontogenesis
myelination
long-chain fatty acid biosynthetic process
synapse organization
endoplasmic reticulum localization
protein localization to plasma membrane
regulation of Golgi organization
microfilament motor activity
actin binding
calcium ion binding
calmodulin binding
ATP binding
Rab GTPase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0