This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06638791_s1
See other CTPS1 GE Assays ›
Gene Symbol:
CTPS1
Gene Name:
CTP synthase 1
Gene Aliases:
CTPS, IMD24
Chromosome Location:
Chr.1: 40979299 - 41012565 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06638791_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTPS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001905.3 NP_001896.2 3 774 65
NR_125440.1 3 774 65
GenBank mRNA AK130549.1 - 2 360 65
AK225899.1 - 3 278 65
AK297386.1 - 3 370 65
AK304817.1 - 3 309 65
BC009408.1 - 3 381 3355881 65
BQ642049.1 - 3 738 65
X52142.1 - 3 305 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CTPS1

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
CTP biosynthetic process
glutamine metabolic process
nucleobase-containing small molecule interconversion
T cell proliferation
B cell proliferation
response to drug
'de novo' CTP biosynthetic process
CTP synthase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0