This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06637086_g1
See other ARHGAP5 GE Assays ›
Gene Symbol:
ARHGAP5
Gene Name:
Rho GTPase activating protein 5
Gene Aliases:
GFI2, RhoGAP5, p190-B, p190BRhoGAP
Chromosome Location:
Chr.14: 32076152 - 32159728 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06637086_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030055.1 NP_001025226.1 2 3943 95
NM_001173.2 NP_001164.2 2 3943 95
XM_005267635.3 XP_005267692.1 2 3912 95
XM_005267636.3 XP_005267693.1 2 4158 95
XM_017021287.1 XP_016876776.1 2 3862 95
XM_017021288.1 XP_016876777.1 2 3912 95
GenBank mRNA AB209751.1 - 1 2541 95
AK292966.1 - 1 3808 95
AY316523.1 - 1 193 95
BC050059.1 - 2 3869 5296634 95
CA945100.1 - 1 231 95
U17032.1 - 2 3927 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP5

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
Rho protein signal transduction
mammary gland development
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activity
GTPase activator activity
protein binding
GTP binding
SH2 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0