This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06635462_s1
See other SH3GLB2 GE Assays ›
Gene Symbol:
SH3GLB2
Gene Name:
SH3 domain containing GRB2 like endophilin B2
Gene Aliases:
PP6569, PP9455, RRIG1
Chromosome Location:
Chr.9: 129007792 - 129028353 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06635462_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SH3GLB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF370426.1 - 1 828 56
AF370433.1 - 1 66 56
AL832592.1 - 1 3184 56

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF370433.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0