This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06633703_s1
See other TIAM2 GE Assays ›
Gene Symbol:
TIAM2
Gene Name:
T-cell lymphoma invasion and metastasis 2
Gene Aliases:
STEF, TIAM-2
Chromosome Location:
Chr.6: 155090289 - 155257723 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06633703_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001010927.2 NP_001010927.1 8 985 76
NM_012454.3 NP_036586.2 21 4233 76
GenBank mRNA AF120323.1 - 17 2203 76
AF120324.1 - 8 961 76
AF195656.1 - 3 282 76
AK123859.1 - 22 4328 76
AK125459.1 - 19 2779 76
AK299723.1 - 8 982 76
AK302860.1 - 17 2144 76
AL122086.1 - 16 2062 76
BC066979.1 - 8 982 76
BC137275.1 - 20 4106 9021272 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TIAM2

Gene Ontology Categories:

Function(s) Process(es)

regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
receptor signaling protein activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0