This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06630959_s1
See other PDE1A GE Assays ›
Gene Symbol:
PDE1A
Gene Name:
phosphodiesterase 1A
Gene Aliases:
CAM-PDE-1A, HCAM-1, HCAM1, HSPDE1A
Chromosome Location:
Chr.2: 182140035 - 182523375 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06630959_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258312.1 NP_001245241.1 1 162 70
XM_011511324.2 XP_011509626.1 1 162 70
XM_017004295.1 XP_016859784.1 1 162 70
XM_017004299.1 XP_016859788.1 1 162 70
GenBank mRNA AF110236.1 - 1 55 70
AK294239.1 - 1 97 70
DA295167.1 - 1 164 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF110236.1, AK294239.1, NM_001258312.1, XM_011511324.2, XM_017004295.1, XM_017004299.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0