This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06629864_s1
See other BCR GE Assays ›
Gene Symbol:
BCR
Gene Name:
BCR, RhoGEF and GTPase activating protein
Gene Aliases:
ALL, BCR1, CML, D22S11, D22S662, PHL
Chromosome Location:
Chr.22: 23180365 - 23318037 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06629864_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004327.3 NP_004318.3 1 1711 59
NM_021574.2 NP_067585.2 1 1711 59
GenBank mRNA AB209991.1 - 1 1460 59
BC066122.1 - 1 1542 59
EU216058.1 - 1 1115 59
EU216059.1 - 1 1115 59
EU216061.1 - 1 1115 59
EU216062.1 - 1 1115 59
EU216063.1 - 1 1115 59
EU216064.1 - 1 1115 59
EU216067.1 - 1 1115 59
EU216069.1 - 1 1115 59
EU216070.1 - 1 1115 59
X02596.1 - 1 1603 59
X06418.1 - 1158 59
X14676.1 - 1 32 59
Y00661.1 - 1 1711 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BCR

Panther Classification:

Molecular Function -

G-protein modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cellular extravasation
protein phosphorylation
signal transduction
brain development
peptidyl-tyrosine phosphorylation
actin cytoskeleton organization
response to lipopolysaccharide
regulation of Rho protein signal transduction
intracellular signal transduction
inner ear morphogenesis
regulation of vascular permeability
negative regulation of neutrophil degranulation
positive regulation of GTPase activity
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
negative regulation of inflammatory response
positive regulation of phagocytosis
neuromuscular process controlling balance
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of blood vessel remodeling
protein serine/threonine kinase activity
protein tyrosine kinase activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
ATP binding
kinase activity
enzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0