This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06627269_s1
See other RBM20 GE Assays ›
Gene Symbol:
RBM20
Gene Name:
RNA binding motif protein 20
Gene Aliases:
-
Chromosome Location:
Chr.10: 110641934 - 110839471 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06627269_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017016103.1 XP_016871592.1 1 809 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017016103.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
heart development
RNA splicing
positive regulation of RNA splicing
nucleotide binding
RNA binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0