This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06613723_g1
See other HIP1R GE Assays ›
Gene Symbol:
HIP1R
Gene Name:
huntingtin interacting protein 1 related
Gene Aliases:
HIP12, HIP3, ILWEQ
Chromosome Location:
Chr.12: 122834498 - 122862961 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06613723_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIP1R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003959.2 NP_003950.1 32 4297 81
XM_011538963.1 XP_011537265.1 32 4600 81
GenBank mRNA AB013384.1 - 26 3646 81
AB014555.1 - 32 4227 81
AK024062.1 - 10 1883 81
BC011354.2 - 1 213 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HIP1R

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
apoptotic process
clathrin-mediated endocytosis
actin binding
protein binding
clathrin binding
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0