Assay Details
Gene Symbol: ASCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK096409.1 | - | 1 | 221 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK096409.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
