This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06599358_s1
See other APOC1 GE Assays ›
Gene Symbol:
APOC1
Gene Name:
apolipoprotein C1
Gene Aliases:
Apo-CI, ApoC-I, apo-CIB, apoC-IB
Chromosome Location:
Chr.19: 44914247 - 44919349 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06599358_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321065.1 NP_001307994.1 1 188 58
NM_001321066.1 NP_001307995.1 1 188 58
GenBank mRNA AF308154.1 - 1 115 58
AF308155.1 - 1 115 58
AK225971.1 - 1 110 58
BC055093.1 - 1 114 6567705 58
DA482306.1 - 1 188 58
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ249921.1 - 1 5041 Chr. 19 - 44909375 - 44914968 [+] on Build GRCh38 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF308154.1, AF308155.1, BC055093.1, NM_001321065.1, NM_001321066.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

apolipoprotein transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
triglyceride metabolic process
cholesterol metabolic process
positive regulation of cholesterol esterification
negative regulation of phosphatidylcholine catabolic process
negative regulation of very-low-density lipoprotein particle clearance
regulation of cholesterol transport
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
plasma lipoprotein particle remodeling
high-density lipoprotein particle remodeling
very-low-density lipoprotein particle assembly
chylomicron remnant clearance
very-low-density lipoprotein particle clearance
lipoprotein metabolic process
positive regulation of catalytic activity
negative regulation of fatty acid biosynthetic process
negative regulation of lipid metabolic process
negative regulation of receptor-mediated endocytosis
negative regulation of lipid catabolic process
negative regulation of lipoprotein lipase activity
phospholipase inhibitor activity
fatty acid binding
phosphatidylcholine binding
lipase inhibitor activity
phosphatidylcholine-sterol O-acyltransferase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0