This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06598187_m1
See other ZMYM4 GE Assays ›
Gene Symbol:
ZMYM4
Gene Name:
zinc finger MYM-type containing 4
Gene Aliases:
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location:
Chr.1: 35268694 - 35421944 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06598187_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMYM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011542423.2 XP_011540725.1 2 - 3 450 90
XM_017002803.1 XP_016858292.1 2 - 3 172 90
GenBank mRNA BC127113.1 - 2 - 3 125 40125110 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC127113.1, XM_011542423.2, XM_017002803.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
cytoskeleton organization
multicellular organism development
regulation of cell morphogenesis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0