This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05649561_s1
See other REXO1 GE Assays ›
Gene Symbol:
REXO1
Gene Name:
RNA exonuclease 1 homolog
Gene Aliases:
ELOABP1, EloA-BP1, REX1, TCEB3BP1
Chromosome Location:
Chr.19: 1815246 - 1848684 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05649561_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: REXO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020695.3 NP_065746.3 16 4362 107
XM_011528144.1 XP_011526446.1 15 4494 107
XM_011528146.2 XP_011526448.1 15 4758 107
XM_017027028.1 XP_016882517.1 15 4467 107
XM_017027029.1 XP_016882518.1 16 4424 107
GenBank mRNA AB032964.1 - 16 4350 107

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene REXO1

Panther Classification:

Molecular Function -

exoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
exonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0