This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05635283_s1
See other DLK1 GE Assays ›
Gene Symbol:
DLK1
Gene Name:
delta like non-canonical Notch ligand 1
Gene Aliases:
DLK, DLK-1, Delta1, FA1, PREF1, Pref-1, ZOG, pG2
Chromosome Location:
Chr.14: 100726865 - 100738224 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05635283_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317172.1 NP_001304101.1 6 2617 90
NM_003836.6 NP_003827.3 5 2836 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DLK1

Panther Classification:

Molecular Function -

membrane-bound signaling molecule

Gene Ontology Categories:

Function(s) Process(es)

cell differentiation
negative regulation of Notch signaling pathway
molecular_function
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0