This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05632608_s1
See other CSRP1 GE Assays ›
Gene Symbol:
CSRP1
Gene Name:
cysteine and glycine rich protein 1
Gene Aliases:
CRP, CRP1, CSRP, CYRP, D1S181E, HEL-141, HEL-S-286
Chromosome Location:
Chr.1: 201483530 - 201507259 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05632608_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSRP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193570.1 NP_001180499.1 6 1818 69
NM_001193571.1 NP_001180500.1 6 1862 69
NM_001193572.1 NP_001180501.1 6 1798 69
NM_004078.2 NP_004069.1 6 1836 69
GenBank mRNA AB209756.1 - 4 5119 69
AK057876.1 - 6 1618 69
AK122812.1 - 6 1631 69
AK126445.1 - 1 3250 69
AK126507.1 - 2 3565 68
AK131508.1 - 4 4090 69
BC004265.1 - 5 1572 69
BC007492.2 - 6 1673 69
BC018806.1 - 6 1691 69
BC032493.1 - 6 1690 5244276 69
BX537488.1 - 2 4691 69
BX641046.1 - 1 2482 69
CA411857.1 - 1 69
CR627407.1 - 2 8356 69
EU668320.1 - 6 1699 69
GQ472204.1 - 5 1572 69
M33146.1 - 6 1678 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CSRP1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

platelet aggregation
zinc ion binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0