Assay Details
Gene Symbol: RNA45S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046235.1 | 1 | 12933 | 75 |
Gene Symbol: RNA28S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003287.2 | 1 | 5009 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
