This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05625588_s1
See other INPP5A GE Assays ›
Gene Symbol:
INPP5A
Gene Name:
inositol polyphosphate-5-phosphatase A
Gene Aliases:
5PTASE
Chromosome Location:
Chr.10: 132537779 - 132783480 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05625588_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: INPP5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017016205.1 XP_016871694.1 12 1646 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

inositol phosphate metabolic process
phosphatidylinositol dephosphorylation
inositol-polyphosphate 5-phosphatase activity
protein binding
PH domain binding
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0