This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05624880_s1
See other SLC16A7 GE Assays ›
Gene Symbol:
SLC16A7
Gene Name:
solute carrier family 16 member 7
Gene Aliases:
MCT2
Chromosome Location:
Chr.12: 59595934 - 59789855 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05624880_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC16A7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270622.1 NP_001257551.1 6 6728 89
NM_001270623.1 NP_001257552.1 6 6748 89
NM_004731.4 NP_004722.2 5 6586 89
NR_073055.1 7 6862 89
NR_073056.1 7 6723 89
XM_005269231.4 XP_005269288.1 5 6522 89
XM_011538989.2 XP_011537291.1 7 6932 89
XM_011538990.2 XP_011537292.1 6 6824 89
XM_011538991.2 XP_011537293.1 6 6622 89
XM_011538992.2 XP_011537294.1 6 7557 89
XM_011538993.2 XP_011537295.1 8 7322 89
XM_011538995.2 XP_011537297.1 8 7317 89
XM_017020224.1 XP_016875713.1 9 7243 89
XM_017020225.1 XP_016875714.1 9 7236 89
XM_017020226.1 XP_016875715.1 9 7371 89
XM_017020227.1 XP_016875716.1 8 6917 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC16A7

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

lactate transmembrane transport
plasma membrane lactate transport
pyruvate transmembrane transport
pyruvate secondary active transmembrane transporter activity
monocarboxylic acid transmembrane transporter activity
lactate transmembrane transporter activity
symporter activity
pyruvate transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0