This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05607937_s1
See other CRCP GE Assays ›
Gene Symbol:
CRCP
Gene Name:
CGRP receptor component
Gene Aliases:
CGRP-RCP, CGRPRCP, RCP, RCP9
Chromosome Location:
Chr.7: 66114818 - 66154568 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05607937_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRCP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040647.1 NP_001035737.1 5 2577 179
NM_001040648.1 NP_001035738.1 3 2445 179
NM_001142414.1 NP_001135886.1 5 2521 179
NM_014478.4 NP_055293.1 6 2676 179
NR_024548.1 7 2798 179
GenBank mRNA AK023295.1 - 2 1497 179
BC052570.1 - 7 3012 179
BX647867.1 - 6 2019 179

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CRCP

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
neuropeptide signaling pathway
positive regulation of type I interferon production
innate immune response
defense response to virus
nucleotide binding
RNA polymerase III activity
calcitonin gene-related peptide receptor activity
DNA-directed RNA polymerase activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0