This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05594545_s1
See other MDM4 GE Assays ›
Gene Symbol:
MDM4
Gene Name:
MDM4, p53 regulator
Gene Aliases:
HDMX, MDMX, MRP1
Chromosome Location:
Chr.1: 204516379 - 204558120 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05594545_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204171.1 NP_001191100.1 10 8115 76
NM_001204172.1 NP_001191101.1 3 7287 76
NM_001278516.1 NP_001265445.1 10 8197 76
NM_001278517.1 NP_001265446.1 8 7971 76
NM_001278518.1 NP_001265447.1 7 7786 76
NM_001278519.1 NP_001265448.1 5 7596 76
NM_002393.4 NP_002384.2 11 8265 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MDM4

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell proliferation
negative regulation of cell proliferation
protein ubiquitination
DNA damage response, signal transduction by p53 class mediator
negative regulation of protein catabolic process
negative regulation of apoptotic process
G0 to G1 transition
protein stabilization
negative regulation of cell cycle arrest
cellular response to hypoxia
regulation of signal transduction by p53 class mediator
protein binding
zinc ion binding
enzyme binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0