This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05592922_s1
See other PDE4D GE Assays ›
Gene Symbol:
PDE4D
Gene Name:
phosphodiesterase 4D
Gene Aliases:
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
Chromosome Location:
Chr.5: 58969038 - 60488098 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05592922_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE4D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248538.4 XP_005248595.1 1 2194 114

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005248538.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
cAMP catabolic process
smooth muscle contraction
aging
regulation of receptor activity
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
cAMP-mediated signaling
neutrophil chemotaxis
regulation of cAMP metabolic process
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-5 production
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
T cell receptor signaling pathway
regulation of ryanodine-sensitive calcium-release channel activity
establishment of endothelial barrier
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
adrenergic receptor signaling pathway involved in positive regulation of heart rate
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
drug binding
enzyme binding
cAMP binding
beta-2 adrenergic receptor binding
ion channel binding
metal ion binding
ATPase binding
scaffold protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0