This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05590177_s1
See other SLC34A2 GE Assays ›
Gene Symbol:
SLC34A2
Gene Name:
solute carrier family 34 member 2
Gene Aliases:
NAPI-3B, NAPI-IIb, NPTIIb
Chromosome Location:
Chr.4: 25655813 - 25678748 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05590177_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC34A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177998.1 NP_001171469.1 13 3305 70
NM_001177999.1 NP_001171470.1 13 3328 70
NM_006424.2 NP_006415.2 13 3308 70
GenBank mRNA AF146796.1 - 13 3260 70
AK075015.1 - 7 2417 70
AK075045.1 - 1 1094 70
BC142704.1 - 13 3254 40148805 70
BC146666.1 - 13 3254 40148124 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC34A2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
phosphate ion transport
cellular phosphate ion homeostasis
phosphate ion transmembrane transport
sodium ion transmembrane transport
response to estrogen
cellular protein metabolic process
sodium-dependent phosphate transport
inorganic phosphate transmembrane transporter activity
sodium:phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
sodium ion binding
phosphate ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0