This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05584196_s1
See other CSRNP1 GE Assays ›
Gene Symbol:
CSRNP1
Gene Name:
cysteine and serine rich nuclear protein 1
Gene Aliases:
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
Chromosome Location:
Chr.3: 39141851 - 39154602 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05584196_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CSRNP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017007048.1 XP_016862537.1 1 1857 112

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017007048.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
apoptotic process
post-embryonic development
positive regulation of transcription from RNA polymerase II promoter
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
molecular_function
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0