This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05581901_s1
See other PBX1 GE Assays ›
Gene Symbol:
PBX1
Gene Name:
PBX homeobox 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 164559360 - 164886047 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05581901_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PBX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011509590.1 XP_011507892.1 10 2622 109
XM_011509592.1 XP_011507894.1 9 2509 109
XM_017001395.1 XP_016856884.1 9 2454 109
XM_017001396.1 XP_016856885.1 8 2341 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
transcription from RNA polymerase II promoter
steroid biosynthetic process
sex differentiation
positive regulation of cell proliferation
anterior/posterior pattern specification
proximal/distal pattern formation
positive regulation of G2/M transition of mitotic cell cycle
regulation of ossification
adrenal gland development
embryonic limb morphogenesis
somatic stem cell population maintenance
embryonic hemopoiesis
negative regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
spleen development
thymus development
embryonic skeletal system development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header