This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05510406_s1
See other PTPRN2 GE Assays ›
Gene Symbol:
PTPRN2
Gene Name:
protein tyrosine phosphatase, receptor type N2
Gene Aliases:
IA-2beta, IAR, ICAAR, PTPRP, R-PTP-N2
Chromosome Location:
Chr.7: 157539052 - 158587802 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05510406_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011516446.1 XP_011514748.1 18 7038 126
XM_017012476.1 XP_016867965.1 18 7084 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protein phosphatase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
neurotransmitter secretion
negative regulation of GTPase activity
peptidyl-tyrosine dephosphorylation
insulin secretion involved in cellular response to glucose stimulus
transmembrane receptor protein tyrosine phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0