This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05484803_s1
See other SRF GE Assays ›
Gene Symbol:
SRF
Gene Name:
serum response factor
Gene Aliases:
MCM1
Chromosome Location:
Chr.6: 43171295 - 43181506 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05484803_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001292001.1 NP_001278930.1 5 755 88
NM_003131.3 NP_003122.1 5 1568 88
GenBank mRNA AB209128.1 - 3 4543 88
AK300467.1 - 5 755 88
BC048211.1 - 5 1404 5808348 88
BC052572.1 - 5 1568 6462845 88
J03161.1 - 5 1564 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SRF

Gene Ontology Categories:

Function(s) Process(es)

patterning of blood vessels
response to hypoxia
mesoderm formation
neuron migration
trophectodermal cell differentiation
heart looping
morphogenesis of an epithelial sheet
cell migration involved in sprouting angiogenesis
positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
transcription from RNA polymerase II promoter
cell-matrix adhesion
heart development
long-term memory
negative regulation of cell proliferation
associative learning
response to toxic substance
response to hormone
epithelial structure maintenance
positive regulation of transcription via serum response element binding
hippocampus development
tangential migration from the subventricular zone to the olfactory bulb
regulation of cell adhesion
platelet activation
platelet formation
negative regulation of cell migration
thyroid gland development
neuron projection development
regulation of water loss via skin
response to cytokine
megakaryocyte development
dorsal aorta morphogenesis
mRNA transcription from RNA polymerase II promoter
stress fiber assembly
skin morphogenesis
positive thymic T cell selection
sarcomere organization
positive regulation of cell differentiation
positive regulation of axon extension
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle contraction
positive regulation of transcription by glucose
muscle cell cellular homeostasis
thymus development
developmental growth
neuron development
erythrocyte development
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of smooth muscle cell differentiation
positive regulation of filopodium assembly
cardiac myofibril assembly
angiogenesis involved in wound healing
hematopoietic stem cell differentiation
positive regulation of transcription initiation from RNA polymerase II promoter
long term synaptic depression
face development
heart trabecula formation
lung morphogenesis
bronchus cartilage development
trachea cartilage development
cardiac vascular smooth muscle cell differentiation
eyelid development in camera-type eye
lung smooth muscle development
bicellular tight junction assembly
cellular response to glucose stimulus
primitive streak formation
epithelial cell-cell adhesion
cellular senescence
negative regulation of beta-amyloid clearance
negative regulation of pri-miRNA transcription from RNA polymerase II promoter
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
serum response element binding
chromatin DNA binding
protein homodimerization activity
histone deacetylase binding
primary miRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0