This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05466429_s1
See other RAD52 GE Assays ›
Gene Symbol:
RAD52
Gene Name:
RAD52 homolog, DNA repair protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 911028 - 991195 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05466429_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAD52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005253720.4 XP_005253777.1 1 300 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005253720.4 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA recombinase assembly
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
DNA double-strand break processing involved in repair via single-strand annealing
protein sumoylation
cellular response to oxidative stress
regulation of nucleotide-excision repair
DNA binding
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0