This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05462497_s1
See other DCPS GE Assays ›
Gene Symbol:
DCPS
Gene Name:
decapping enzyme, scavenger
Gene Aliases:
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015
Chromosome Location:
Chr.11: 126303301 - 126345754 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05462497_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCPS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017017612.1 XP_016873101.1 1 127 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017017612.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
deadenylation-dependent decapping of nuclear-transcribed mRNA
cellular response to menadione
negative regulation of programmed cell death
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
mRNA cis splicing, via spliceosome
RNA phosphodiester bond hydrolysis, exonucleolytic
RNA 7-methylguanosine cap binding
exoribonuclease activity
protein binding
m7G(5')pppN diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0