This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05449771_g1
See other GNL3 GE Assays ›
Gene Symbol:
GNL3
Gene Name:
G protein nucleolar 3
Gene Aliases:
C77032, E2IG3, NNP47, NS
Chromosome Location:
Chr.3: 52685920 - 52694497 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05449771_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014366.4 NP_055181.3 15 1963 120
NM_206825.1 NP_996561.1 15 2060 120
NM_206826.1 NP_996562.1 15 1913 120
GenBank mRNA AF191018.1 - 15 1960 120
AK027514.1 - 15 1964 120
AK027516.1 - 15 1813 120
BC001024.2 - 15 1812 3342771 120

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GNL3

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
positive regulation of telomere maintenance
positive regulation of protein sumoylation
regulation of cell proliferation
ribosome biogenesis
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
positive regulation of protein localization to chromosome, telomeric region
GTPase activity
protein binding
GTP binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0