This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05446698_s1
See other GRID2 GE Assays ›
Gene Symbol:
GRID2
Gene Name:
glutamate ionotropic receptor delta type subunit 2
Gene Aliases:
GluD2, SCAR18
Chromosome Location:
Chr.4: 92304249 - 93810330 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05446698_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRID2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011531893.2 XP_011530195.2 1 5763 100
XM_017008118.1 XP_016863607.1 1 5763 100
XM_017008120.1 XP_016863609.1 1 5763 100
XM_017008121.1 XP_016863610.1 1 5763 100
XM_017008127.1 XP_016863616.1 1 5763 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011531893.2, XM_017008118.1, XM_017008120.1, XM_017008121.1, XM_017008127.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transport
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
glutamate receptor signaling pathway
regulation of neuron projection development
cerebellar granule cell differentiation
ion transmembrane transport
cellular protein localization
ionotropic glutamate receptor signaling pathway
synaptic transmission, glutamatergic
regulation of neuron apoptotic process
excitatory postsynaptic potential
prepulse inhibition
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
PDZ domain binding
scaffold protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0