This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05422826_s1
See other SLC6A6 GE Assays ›
Gene Symbol:
SLC6A6
Gene Name:
solute carrier family 6 member 6
Gene Aliases:
TAUT
Chromosome Location:
Chr.3: 14402576 - 14489349 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05422826_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC6A6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134367.3 NP_001127839.2 15 4624 82
NM_003043.5 NP_003034.2 15 4628 82
NR_103507.2 14 4503 82
XM_006713307.2 XP_006713370.1 14 4432 82
XM_011534030.2 XP_011532332.1 14 4385 82
XM_011534031.2 XP_011532333.1 16 4756 82
GenBank mRNA AB209172.1 - 14 4428 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC6A6

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
cellular amino acid metabolic process
transport
neurotransmitter transport
amino acid transport
taurine transport
neurotransmitter:sodium symporter activity
taurine:sodium symporter activity
amino acid transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0