This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05412665_s1
See other IL6R GE Assays ›
Gene Symbol:
IL6R
Gene Name:
interleukin 6 receptor
Gene Aliases:
CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
Chromosome Location:
Chr.1: 154405193 - 154469450 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05412665_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL6R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000565.3 NP_000556.1 10 5436 109
NM_181359.2 NP_852004.1 9 5342 109
XM_005245139.1 XP_005245196.1 8 5159 109
XM_006711298.1 XP_006711361.1 11 5443 109
XM_017001199.1 XP_016856688.1 12 5542 109
XM_017001200.1 XP_016856689.1 11 5494 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL6R

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

hepatic immune response
monocyte chemotaxis
positive regulation of leukocyte chemotaxis
acute-phase response
positive regulation of cell proliferation
cytokine-mediated signaling pathway
endocrine pancreas development
positive regulation of chemokine production
positive regulation of interleukin-6 production
response to cytokine
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of osteoblast differentiation
positive regulation of smooth muscle cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
interleukin-6-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
extrinsic apoptotic signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-6 receptor binding
protein binding
enzyme binding
interleukin-6 binding
protein homodimerization activity
ciliary neurotrophic factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0