This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05394227_g1
See other KCNJ6 GE Assays ›
Gene Symbol:
KCNJ6
Gene Name:
potassium voltage-gated channel subfamily J member 6
Gene Aliases:
BIR1, GIRK-2, GIRK2, KATP-2, KATP2, KCNJ7, KIR3.2, KPLBS, hiGIRK2
Chromosome Location:
Chr.21: 37624223 - 37916438 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05394227_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KCNJ6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002240.4 NP_002231.1 1 269 100
GenBank mRNA D87327.1 - 1 190 100
DR000377.1 - 1 276 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the D87327.1, NM_002240.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KCNJ6

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
potassium ion import
regulation of ion transmembrane transport
inward rectifier potassium channel activity
protein binding
G-protein activated inward rectifier potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0