This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05381682_s1
See other NRP2 GE Assays ›
Gene Symbol:
NRP2
Gene Name:
neuropilin 2
Gene Aliases:
NP2, NPN2, PRO2714, VEGF165R2
Chromosome Location:
Chr.2: 205682450 - 205798133 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05381682_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003872.2 NP_003863.2 1 545 95
NM_018534.3 NP_061004.3 1 545 95
NM_201264.1 NP_957716.1 1 545 95
NM_201266.1 NP_957718.1 1 545 95
NM_201267.1 NP_957719.1 1 545 95
NM_201279.1 NP_958436.1 1 545 95
XM_017005187.1 XP_016860676.1 1 595 95
XM_017005188.1 XP_016860677.1 1 594 95
GenBank mRNA AK130198.1 - 2 531 95
AL833606.1 - 1 545 95
BX537423.1 - 1 544 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BX537423.1, NM_003872.2, NM_018534.3, NM_201264.1, NM_201266.1, NM_201267.1, NM_201279.1, XM_017005187.1, XM_017005188.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NRP2

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
positive regulation of endothelial cell proliferation
cell adhesion
axon guidance
positive regulation of endothelial cell migration
nerve development
vascular endothelial growth factor signaling pathway
vascular endothelial growth factor receptor signaling pathway
axon extension involved in axon guidance
sympathetic ganglion development
sympathetic neuron projection extension
sympathetic neuron projection guidance
semaphorin-plexin signaling pathway involved in neuron projection guidance
receptor activity
vascular endothelial growth factor-activated receptor activity
heparin binding
semaphorin receptor activity
growth factor binding
cytokine binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0