This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05380290_s1
See other PDK1 GE Assays ›
Gene Symbol:
PDK1
Gene Name:
pyruvate dehydrogenase kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 172555373 - 172672693 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05380290_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278549.1 NP_001265478.1 12 2072 69
NM_002610.4 NP_002601.1 11 2012 69
XM_006712595.3 XP_006712658.1 9 1337 69
XM_011511343.2 XP_011509645.1 12 1834 69
XM_011511344.2 XP_011509646.1 12 1719 69
XM_011511345.2 XP_011509647.1 11 1774 69
XM_011511347.2 XP_011509649.1 12 1719 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDK1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
protein phosphorylation
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
hypoxia-inducible factor-1alpha signaling pathway
mitophagy in response to mitochondrial depolarization
protein kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0