Assay Details
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL122104.1 | - | 2 | 1058 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL122104.1 | - | 2 | 1058 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |