This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05342578_s1
See other PHLPP1 GE Assays ›
Gene Symbol:
PHLPP1
Gene Name:
PH domain and leucine rich repeat protein phosphatase 1
Gene Aliases:
PHLPP, PLEKHE1, PPM3A, SCOP
Chromosome Location:
Chr.18: 62715439 - 62980443 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05342578_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHLPP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194449.3 NP_919431.2 17 6192 88
GenBank mRNA AB011178.2 - 17 4905 88
BC010706.1 - 4 2427 88
BC014927.2 - 17 4479 3916044 88
BC047653.1 - 6 2766 88
BC063519.1 - 5 2637 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHLPP1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of T cell anergy
protein dephosphorylation
apoptotic process
entrainment of circadian clock
regulation of apoptotic process
regulation of MAPK cascade
regulation of JNK cascade
negative regulation of protein kinase B signaling
regulation of p38MAPK cascade
phosphoprotein phosphatase activity
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0