This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05341552_s1
See other SPTBN2 GE Assays ›
Gene Symbol:
SPTBN2
Gene Name:
spectrin beta, non-erythrocytic 2
Gene Aliases:
GTRAP41, SCA5, SCAR14
Chromosome Location:
Chr.11: 66682496 - 66729361 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05341552_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPTBN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017018175.1 XP_016873664.1 1 182 92
XM_017018177.1 XP_016873666.1 1 182 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017018175.1, XM_017018177.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
ER to Golgi vesicle-mediated transport
cytoskeleton organization
axon guidance
synapse assembly
vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
cerebellar Purkinje cell layer morphogenesis
adult behavior
multicellular organism growth
positive regulation of GTPase activity
actin filament capping
cellular response to ethanol
cell-cell adhesion
actin binding
Ras guanyl-nucleotide exchange factor activity
structural constituent of cytoskeleton
phospholipid binding
cadherin binding involved in cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0