This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05330575_s1
See other PITPNM3 GE Assays ›
Gene Symbol:
PITPNM3
Gene Name:
PITPNM family member 3
Gene Aliases:
ACKR6, CORD5, NIR1, RDGBA3
Chromosome Location:
Chr.17: 6451263 - 6556557 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05330575_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PITPNM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011524016.2 XP_011522318.1 18 3791 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

phospholipid transport
phosphatidylinositol metabolic process
calcium ion binding
protein binding
lipid binding
phosphatidylinositol transporter activity
receptor tyrosine kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0