This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05323914_s1
See other PPP1R9B GE Assays ›
Gene Symbol:
PPP1R9B
Gene Name:
protein phosphatase 1 regulatory subunit 9B
Gene Aliases:
PPP1R6, PPP1R9, SPINO, Spn
Chromosome Location:
Chr.17: 50133735 - 50151027 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05323914_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R9B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011525381.2 XP_011523683.1 1 314 101

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011525381.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth by extracellular stimulus
actin filament organization
cell cycle arrest
regulation of exit from mitosis
RNA splicing
cell migration
calcium-mediated signaling
negative regulation of cell growth
neuron projection development
regulation of cell proliferation
negative regulation of catalytic activity
filopodium assembly
modulation of synaptic transmission
cellular response to morphine
regulation of opioid receptor signaling pathway
protein phosphatase inhibitor activity
protein binding
protein phosphatase 1 binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0