This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05316003_s1
See other SETD1A GE Assays ›
Gene Symbol:
SETD1A
Gene Name:
SET domain containing 1A
Gene Aliases:
KMT2F, Set1, Set1A
Chromosome Location:
Chr.16: 30956618 - 30984664 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05316003_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SETD1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017023909.1 XP_016879398.1 1 356 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017023909.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
nucleotide binding
RNA binding
protein binding
beta-catenin binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0