This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05289949_s1
See other IGHM GE Assays ›
Gene Symbol:
IGHM
Gene Name:
immunoglobulin heavy constant mu
Gene Aliases:
AGM1, MU, VH
Chromosome Location:
Chr.14: 105851966 - 105856217 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05289949_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGHM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC001872.1 - 6 1273 139
BC002963.1 - 6 1253 139
BC006180.1 - 6 1273 139
BC009851.2 - 6 1234 139
BC011857.2 - 7 1253 139
BC015760.1 - 6 1305 139
BC017356.2 - 6 1270 139
BC019235.2 - 6 1272 139
BC020240.1 - 6 1301 139
BC073758.1 - 6 1342 139
BC073767.1 - 6 1261 139
BC089412.1 - 5 1267 139
BX161420.1 - 3 1271 139
X17115.1 - 7 1345 139
X58529.1 - 4 1458 139
X67292.1 - 3 814 139
X67301.1 - 3 814 139
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC013877.1 - 2143 Chr. - - [-] on Build 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0