This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05273190_s1
See other ATP8A2 GE Assays ›
Gene Symbol:
ATP8A2
Gene Name:
ATPase phospholipid transporting 8A2
Gene Aliases:
ATP, ATPIB, CAMRQ4, IB, ML-1
Chromosome Location:
Chr.13: 25372011 - 26025851 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05273190_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP8A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011535107.2 XP_011533409.1 34 4232 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

involuntary skeletal muscle contraction
axonogenesis
aging
negative regulation of cell proliferation
retina layer formation
positive regulation of neuron projection development
response to auditory stimulus
positive regulation of multicellular organism growth
inner ear morphogenesis
eating behavior
skin development
phospholipid translocation
Golgi vesicle budding
neuron development
neuromuscular process controlling posture
detection of light stimulus involved in visual perception
neurofilament cytoskeleton organization
positive regulation of phospholipid translocation
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0